Assay Details
| Gene Symbol: 4E-T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272149.2 | NP_001259078.1 | 2 - 3 | 1507 | 122 | |
| NM_166796.2 | NP_726618.3 | 3 - 4 | 1564 | 122 | ||
| NM_166797.2 | NP_726619.2 | 3 - 4 | 1539 | 122 | ||
| NM_166798.2 | NP_726620.2 | 2 - 3 | 1729 | 122 | ||
| GenBank mRNA | BT001473.1 | - | 2 - 3 | 1729 | 122 | |
More Information
Set Membership: |
|

4E-T
Genomic Map