Assay Details
| Gene Symbol: 4E-T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166796.2 | NP_726618.3 | 6 - 7 | 2274 | 96 | |
| NM_166797.2 | NP_726619.2 | 6 - 7 | 2249 | 96 | ||
| NM_166798.2 | NP_726620.2 | 5 - 6 | 2439 | 96 | ||
| GenBank mRNA | BT001473.1 | - | 5 - 6 | 2439 | 96 | |
More Information
Set Membership: |
|

4E-T
Genomic Map