Assay Details
| Gene Symbol: 4E-T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272149.2 | NP_001259078.1 | 6 - 7 | 2517 | 70 | |
| NM_166796.2 | NP_726618.3 | 7 - 8 | 2667 | 70 | ||
| NM_166797.2 | NP_726619.2 | 7 - 8 | 2642 | 70 | ||
| NM_166798.2 | NP_726620.2 | 6 - 7 | 2832 | 70 | ||
| GenBank mRNA | BT001473.1 | - | 6 - 7 | 2832 | 70 | |
More Information
Set Membership: |
|

4E-T
Genomic Map