Assay Details
| Gene Symbol: CaMKII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014696.2 | NP_001014696.1 | 6 - 7 | 892 | 72 | |
| NM_001169360.2 | NP_001162831.1 | 6 - 7 | 1371 | 72 | ||
| NM_001169361.2 | NP_001162832.1 | 7 - 8 | 788 | 72 | ||
| NM_001169362.2 | NP_001162833.1 | 6 - 7 | 722 | 72 | ||
| NM_001272153.1 | NP_001259082.1 | 6 - 7 | 892 | 72 | ||
| NM_001272154.1 | NP_001259083.1 | 6 - 7 | 892 | 72 | ||
| NM_001272155.1 | NP_001259084.1 | 6 - 7 | 892 | 72 | ||
| NM_079896.4 | NP_524635.3 | 6 - 7 | 892 | 72 | ||
| NM_166810.5 | NP_726633.2 | 6 - 7 | 722 | 72 | ||
| NM_166811.2 | NP_726634.1 | 6 - 7 | 806 | 72 | ||
| NM_166812.3 | NP_726635.2 | 6 - 7 | 892 | 72 | ||
| NM_166813.3 | NP_726636.2 | 6 - 7 | 892 | 72 | ||
| GenBank mRNA | BT050453.1 | - | 6 - 7 | 1310 | 72 | |
| M74583.1 | - | 6 - 7 | 846 | 72 | ||
More Information
Set Membership: |
|

CaMKII
Genomic Map