Assay Details
| Gene Symbol: CaMKII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014696.2 | NP_001014696.1 | 10 - 11 | 1329 | 151 | |
| NM_001169361.2 | NP_001162832.1 | 11 - 12 | 1225 | 151 | ||
| NM_001169362.2 | NP_001162833.1 | 10 - 11 | 1159 | 151 | ||
| NM_166811.2 | NP_726634.1 | 10 - 11 | 1243 | 151 | ||
| NM_166812.3 | NP_726635.2 | 10 - 11 | 1329 | 151 | ||
| NM_166813.3 | NP_726636.2 | 10 - 11 | 1329 | 151 | ||
More Information
Set Membership: |
|

CaMKII
Genomic Map