Assay Details
| Gene Symbol: sv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014697.2 | NP_001014697.1 | 3 - 4 | 595 | 129 | |
| NM_001014698.3 | NP_001014698.1 | 3 - 4 | 781 | 129 | ||
| NM_001014699.3 | NP_001014699.1 | 3 - 4 | 781 | 129 | ||
| NM_001014701.2 | NP_001014701.1 | 3 - 4 | 781 | 129 | ||
| NM_079894.4 | NP_524633.3 | 3 - 4 | 781 | 129 | ||
| NM_166822.4 | NP_726645.3 | 3 - 4 | 781 | 129 | ||
| GenBank mRNA | AF010256.1 | - | 3 - 4 | 761 | 129 | |
More Information
Set Membership: |
|

sv
Genomic Map