Assay Details
| Gene Symbol: pho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079891.3 | NP_524630.1 | 3 - 4 | 638 | 123 | |
| NM_166827.2 | NP_726651.1 | 3 - 4 | 416 | 123 | ||
| GenBank mRNA | AF034212.1 | - | 3 - 4 | 569 | 123 | |
| AY071143.1 | - | 3 - 4 | 655 | 123 | ||
More Information
Set Membership: |
|

pho
Genomic Map