Assay Details
| Gene Symbol: cin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057682.4 | NP_477030.1 | 2 - 3 | 907 | 88 | |
| NM_166835.4 | NP_726659.1 | 2 - 3 | 907 | 88 | ||
| GenBank mRNA | AY069078.1 | - | 3 - 4 | 1275 | 88 | |
More Information
Set Membership: |
|

cin
Genomic Map