Assay Details
| Gene Symbol: cin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057682.4 | NP_477030.1 | 4 - 5 | 1518 | 81 | |
| NM_166835.4 | NP_726659.1 | 4 - 5 | 1518 | 81 | ||
| GenBank mRNA | AY069078.1 | - | 5 - 6 | 1886 | 81 | |
More Information
Set Membership: |
|

cin
Genomic Map