Assay Details
| Gene Symbol: csw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272245.1 | NP_001259174.1 | 5 - 6 | 1360 | 66 | |
| NM_057782.5 | NP_477130.1 | 5 - 6 | 838 | 66 | ||
| NM_057783.3 | NP_477131.1 | 3 - 4 | 1825 | 66 | ||
| NM_166928.2 | NP_726793.1 | 3 - 4 | 384 | 66 | ||
| GenBank mRNA | BT001484.1 | - | 5 - 6 | 772 | 66 | |
| BT149891.1 | - | 3 - 4 | 1629 | 66 | ||
| M94730.1 | - | 5 - 6 | 807 | 66 | ||
More Information
Set Membership: |
|

csw
Genomic Map