Assay Details
| Gene Symbol: csw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272245.1 | NP_001259174.1 | 8 - 9 | 2506 | 81 | |
| NM_057782.5 | NP_477130.1 | 8 - 9 | 1984 | 81 | ||
| NM_057783.3 | NP_477131.1 | 6 - 7 | 2971 | 81 | ||
| NM_166928.2 | NP_726793.1 | 6 - 7 | 1530 | 81 | ||
| GenBank mRNA | BT001484.1 | - | 8 - 9 | 1918 | 81 | |
| BT149891.1 | - | 6 - 7 | 2775 | 81 | ||
| M94730.1 | - | 8 - 9 | 1953 | 81 | ||
More Information
Set Membership: |
|

csw
Genomic Map