Assay Details
| Gene Symbol: csw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272245.1 | NP_001259174.1 | 9 - 10 | 2706 | 73 | |
| NM_057782.5 | NP_477130.1 | 9 - 10 | 2184 | 73 | ||
| NM_057783.3 | NP_477131.1 | 7 - 8 | 3171 | 73 | ||
| NM_166928.2 | NP_726793.1 | 7 - 8 | 1730 | 73 | ||
| GenBank mRNA | BT001484.1 | - | 9 - 10 | 2118 | 73 | |
| BT149891.1 | - | 7 - 8 | 2975 | 73 | ||
| M94730.1 | - | 9 - 10 | 2153 | 73 | ||
More Information
Set Membership: |
|

csw
Genomic Map