Assay Details
| Gene Symbol: trol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031862.4 | NP_001027033.3 | 28 - 29 | 9492 | 75 | |
| NM_001031863.5 | NP_001027034.2 | 35 - 36 | 10163 | 75 | ||
| NM_001031864.4 | NP_001027035.3 | 27 - 28 | 9132 | 75 | ||
| NM_001031866.4 | NP_001027037.3 | 30 - 31 | 9921 | 75 | ||
| NM_001103399.4 | NP_001096869.3 | 27 - 28 | 9438 | 75 | ||
| NM_001169177.4 | NP_001162648.3 | 36 - 37 | 10704 | 75 | ||
| NM_001258562.3 | NP_001245491.2 | 37 - 38 | 10824 | 75 | ||
| NM_001258563.3 | NP_001245492.2 | 24 - 25 | 8655 | 75 | ||
| NM_001258564.3 | NP_001245493.2 | 36 - 37 | 10773 | 75 | ||
| NM_001258565.3 | NP_001245494.2 | 26 - 27 | 8970 | 75 | ||
| NM_001258566.4 | NP_001245495.2 | 35 - 36 | 10006 | 75 | ||
| NM_001258568.3 | NP_001245497.2 | 27 - 28 | 9372 | 75 | ||
| NM_001258569.3 | NP_001245498.2 | 25 - 26 | 8889 | 75 | ||
| NM_001258570.3 | NP_001245499.2 | 33 - 34 | 10368 | 75 | ||
| NM_001258571.3 | NP_001245500.2 | 30 - 31 | 9921 | 75 | ||
| NM_001297893.2 | NP_001284822.1 | 25 - 26 | 8886 | 75 | ||
| NM_001297894.2 | NP_001284823.1 | 27 - 28 | 9390 | 75 | ||
| NM_001297896.2 | NP_001284825.1 | 31 - 32 | 10080 | 75 | ||
More Information
Set Membership: |
|

trol
Genomic Map