Assay Details
| Gene Symbol: crm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057757.4 | NP_477105.2 | 3 - 4 | 2398 | 73 | |
| NM_166951.2 | NP_726829.1 | 4 - 5 | 2455 | 73 | ||
| GenBank mRNA | AY051767.1 | - | 4 - 5 | 2459 | 73 | |
More Information
Set Membership: |
|

crm
Genomic Map