Assay Details
| Gene Symbol: crm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057757.4 | NP_477105.2 | 4 - 5 | 2765 | 76 | |
| NM_166951.2 | NP_726829.1 | 5 - 6 | 2822 | 76 | ||
| GenBank mRNA | AY051767.1 | - | 5 - 6 | 2826 | 76 | |
More Information
Set Membership: |
|

crm
Genomic Map