Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 12 - 13 | 4714 | 99 | |
| NM_001272273.1 | NP_001259202.1 | 9 - 10 | 2883 | 99 | ||
| NM_001272274.1 | NP_001259203.1 | 11 - 12 | 4011 | 99 | ||
| NM_080322.3 | NP_525061.2 | 6 - 7 | 1430 | 99 | ||
| NM_166959.2 | NP_726846.1 | 12 - 13 | 4131 | 99 | ||
| NM_166960.3 | NP_726847.2 | 11 - 12 | 3088 | 99 | ||
| NM_166961.2 | NP_726849.1 | 10 - 11 | 2829 | 99 | ||
| NM_166962.2 | NP_726850.1 | 9 - 10 | 2787 | 99 | ||
| NM_166963.2 | NP_726851.3 | 11 - 12 | 3246 | 99 | ||
| NM_166964.2 | NP_726852.2 | 10 - 11 | 2823 | 99 | ||
| NM_166965.2 | NP_726853.2 | 10 - 11 | 2826 | 99 | ||
| NM_166966.3 | NP_726854.2 | 8 - 9 | 2076 | 99 | ||
| NM_166967.2 | NP_726855.1 | 8 - 9 | 1836 | 99 | ||
| NM_166968.3 | NP_726856.1 | 11 - 12 | 5683 | 99 | ||
| NM_166969.2 | NP_726857.1 | 8 - 9 | 1381 | 99 | ||
| NM_166970.2 | NP_726858.2 | 7 - 8 | 1599 | 99 | ||
| NM_166971.2 | NP_726859.2 | 4 - 5 | 1162 | 99 | ||
| GenBank mRNA | AY119511.1 | - | 4 - 5 | 1162 | 99 | |
| BT133288.1 | - | 11 - 12 | 3086 | 99 | ||
More Information
Set Membership: |
|

dnc
Genomic Map