Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 13 - 14 | 4833 | 148 | |
| NM_001272273.1 | NP_001259202.1 | 10 - 11 | 3002 | 148 | ||
| NM_001272274.1 | NP_001259203.1 | 12 - 13 | 4130 | 148 | ||
| NM_080322.3 | NP_525061.2 | 7 - 8 | 1549 | 148 | ||
| NM_166959.2 | NP_726846.1 | 13 - 14 | 4250 | 148 | ||
| NM_166960.3 | NP_726847.2 | 12 - 13 | 3207 | 148 | ||
| NM_166961.2 | NP_726849.1 | 11 - 12 | 2948 | 148 | ||
| NM_166962.2 | NP_726850.1 | 10 - 11 | 2906 | 148 | ||
| NM_166963.2 | NP_726851.3 | 12 - 13 | 3365 | 148 | ||
| NM_166964.2 | NP_726852.2 | 11 - 12 | 2942 | 148 | ||
| NM_166965.2 | NP_726853.2 | 11 - 12 | 2945 | 148 | ||
| NM_166966.3 | NP_726854.2 | 9 - 10 | 2195 | 148 | ||
| NM_166967.2 | NP_726855.1 | 9 - 10 | 1955 | 148 | ||
| NM_166968.3 | NP_726856.1 | 12 - 13 | 5802 | 148 | ||
| NM_166969.2 | NP_726857.1 | 9 - 10 | 1500 | 148 | ||
| NM_166970.2 | NP_726858.2 | 8 - 9 | 1718 | 148 | ||
| NM_166971.2 | NP_726859.2 | 5 - 6 | 1281 | 148 | ||
| GenBank mRNA | AY119511.1 | - | 5 - 6 | 1281 | 148 | |
| BT133288.1 | - | 12 - 13 | 3205 | 148 | ||
More Information
Set Membership: |
|

dnc
Genomic Map