Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 15 - 16 | 5362 | 58 | |
| NM_001272273.1 | NP_001259202.1 | 12 - 13 | 3531 | 58 | ||
| NM_001272274.1 | NP_001259203.1 | 14 - 15 | 4659 | 58 | ||
| NM_080322.3 | NP_525061.2 | 9 - 10 | 2078 | 58 | ||
| NM_166959.2 | NP_726846.1 | 15 - 16 | 4779 | 58 | ||
| NM_166960.3 | NP_726847.2 | 14 - 15 | 3736 | 58 | ||
| NM_166961.2 | NP_726849.1 | 13 - 14 | 3477 | 58 | ||
| NM_166962.2 | NP_726850.1 | 12 - 13 | 3435 | 58 | ||
| NM_166963.2 | NP_726851.3 | 14 - 15 | 3894 | 58 | ||
| NM_166964.2 | NP_726852.2 | 13 - 14 | 3471 | 58 | ||
| NM_166965.2 | NP_726853.2 | 13 - 14 | 3474 | 58 | ||
| NM_166966.3 | NP_726854.2 | 11 - 12 | 2724 | 58 | ||
| NM_166967.2 | NP_726855.1 | 11 - 12 | 2484 | 58 | ||
| NM_166968.3 | NP_726856.1 | 14 - 15 | 6331 | 58 | ||
| NM_166969.2 | NP_726857.1 | 11 - 12 | 2029 | 58 | ||
| NM_166970.2 | NP_726858.2 | 10 - 11 | 2247 | 58 | ||
| NM_166971.2 | NP_726859.2 | 7 - 8 | 1810 | 58 | ||
| GenBank mRNA | AY119511.1 | - | 7 - 8 | 1810 | 58 | |
| BT133288.1 | - | 14 - 15 | 3734 | 58 | ||
More Information
Set Membership: |
|

dnc
Genomic Map