Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272274.1 | NP_001259203.1 | 3 - 4 | 2412 | 78 | |
| NM_166959.2 | NP_726846.1 | 4 - 5 | 2532 | 78 | ||
| NM_166960.3 | NP_726847.2 | 3 - 4 | 1489 | 78 | ||
| NM_166963.2 | NP_726851.3 | 3 - 4 | 1647 | 78 | ||
| GenBank mRNA | BT133288.1 | - | 3 - 4 | 1487 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001272274.1, NM_166963.2 transcripts.
Set Membership: |
|

dnc
Genomic Map