Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272274.1 | NP_001259203.1 | 4 - 5 | 2802 | 62 | |
| NM_166959.2 | NP_726846.1 | 5 - 6 | 2922 | 62 | ||
| NM_166960.3 | NP_726847.2 | 4 - 5 | 1879 | 62 | ||
| NM_166963.2 | NP_726851.3 | 4 - 5 | 2037 | 62 | ||
| GenBank mRNA | BT133288.1 | - | 4 - 5 | 1877 | 62 | |
More Information
Set Membership: |
|

dnc
Genomic Map