Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 7 - 8 | 3721 | 81 | |
| NM_001272273.1 | NP_001259202.1 | 4 - 5 | 1890 | 81 | ||
| NM_001272274.1 | NP_001259203.1 | 6 - 7 | 3018 | 81 | ||
| NM_166959.2 | NP_726846.1 | 7 - 8 | 3138 | 81 | ||
| NM_166960.3 | NP_726847.2 | 6 - 7 | 2095 | 81 | ||
| NM_166961.2 | NP_726849.1 | 5 - 6 | 1836 | 81 | ||
| NM_166963.2 | NP_726851.3 | 6 - 7 | 2253 | 81 | ||
| NM_166964.2 | NP_726852.2 | 5 - 6 | 1836 | 81 | ||
| NM_166965.2 | NP_726853.2 | 5 - 6 | 1833 | 81 | ||
| NM_166966.3 | NP_726854.2 | 3 - 4 | 1083 | 81 | ||
| NM_166967.2 | NP_726855.1 | 3 - 4 | 843 | 81 | ||
| NM_166969.2 | NP_726857.1 | 3 - 4 | 388 | 81 | ||
| GenBank mRNA | BT133288.1 | - | 6 - 7 | 2093 | 81 | |
More Information
Set Membership: |
|

dnc
Genomic Map