Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 8 - 9 | 3828 | 60 | |
| NM_001272273.1 | NP_001259202.1 | 5 - 6 | 1997 | 60 | ||
| NM_001272274.1 | NP_001259203.1 | 7 - 8 | 3125 | 60 | ||
| NM_080322.3 | NP_525061.2 | 2 - 3 | 544 | 60 | ||
| NM_166959.2 | NP_726846.1 | 8 - 9 | 3245 | 60 | ||
| NM_166960.3 | NP_726847.2 | 7 - 8 | 2202 | 60 | ||
| NM_166961.2 | NP_726849.1 | 6 - 7 | 1943 | 60 | ||
| NM_166962.2 | NP_726850.1 | 5 - 6 | 1901 | 60 | ||
| NM_166963.2 | NP_726851.3 | 7 - 8 | 2360 | 60 | ||
| NM_166965.2 | NP_726853.2 | 6 - 7 | 1940 | 60 | ||
| NM_166966.3 | NP_726854.2 | 4 - 5 | 1190 | 60 | ||
| NM_166967.2 | NP_726855.1 | 4 - 5 | 950 | 60 | ||
| NM_166968.3 | NP_726856.1 | 7 - 8 | 4797 | 60 | ||
| NM_166969.2 | NP_726857.1 | 4 - 5 | 495 | 60 | ||
| NM_166970.2 | NP_726858.2 | 3 - 4 | 713 | 60 | ||
| GenBank mRNA | BT133288.1 | - | 7 - 8 | 2200 | 60 | |
More Information
Set Membership: |
|

dnc
Genomic Map