Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 9 - 10 | 4014 | 62 | |
| NM_001272273.1 | NP_001259202.1 | 6 - 7 | 2183 | 62 | ||
| NM_001272274.1 | NP_001259203.1 | 8 - 9 | 3311 | 62 | ||
| NM_080322.3 | NP_525061.2 | 3 - 4 | 730 | 62 | ||
| NM_166959.2 | NP_726846.1 | 9 - 10 | 3431 | 62 | ||
| NM_166960.3 | NP_726847.2 | 8 - 9 | 2388 | 62 | ||
| NM_166961.2 | NP_726849.1 | 7 - 8 | 2129 | 62 | ||
| NM_166962.2 | NP_726850.1 | 6 - 7 | 2087 | 62 | ||
| NM_166963.2 | NP_726851.3 | 8 - 9 | 2546 | 62 | ||
| NM_166964.2 | NP_726852.2 | 7 - 8 | 2123 | 62 | ||
| NM_166965.2 | NP_726853.2 | 7 - 8 | 2126 | 62 | ||
| NM_166966.3 | NP_726854.2 | 5 - 6 | 1376 | 62 | ||
| NM_166967.2 | NP_726855.1 | 5 - 6 | 1136 | 62 | ||
| NM_166968.3 | NP_726856.1 | 8 - 9 | 4983 | 62 | ||
| NM_166969.2 | NP_726857.1 | 5 - 6 | 681 | 62 | ||
| NM_166970.2 | NP_726858.2 | 4 - 5 | 899 | 62 | ||
| NM_166971.2 | NP_726859.2 | 1 - 2 | 462 | 62 | ||
| GenBank mRNA | AY119511.1 | - | 1 - 2 | 462 | 62 | |
| BT133288.1 | - | 8 - 9 | 2386 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY119511.1, NM_166971.2 transcripts.
Set Membership: |
|

dnc
Genomic Map