Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272272.1 | NP_001259201.1 | 10 - 11 | 4113 | 85 | |
| NM_001272273.1 | NP_001259202.1 | 7 - 8 | 2282 | 85 | ||
| NM_001272274.1 | NP_001259203.1 | 9 - 10 | 3410 | 85 | ||
| NM_080322.3 | NP_525061.2 | 4 - 5 | 829 | 85 | ||
| NM_166959.2 | NP_726846.1 | 10 - 11 | 3530 | 85 | ||
| NM_166960.3 | NP_726847.2 | 9 - 10 | 2487 | 85 | ||
| NM_166961.2 | NP_726849.1 | 8 - 9 | 2228 | 85 | ||
| NM_166962.2 | NP_726850.1 | 7 - 8 | 2186 | 85 | ||
| NM_166963.2 | NP_726851.3 | 9 - 10 | 2645 | 85 | ||
| NM_166964.2 | NP_726852.2 | 8 - 9 | 2222 | 85 | ||
| NM_166965.2 | NP_726853.2 | 8 - 9 | 2225 | 85 | ||
| NM_166966.3 | NP_726854.2 | 6 - 7 | 1475 | 85 | ||
| NM_166967.2 | NP_726855.1 | 6 - 7 | 1235 | 85 | ||
| NM_166968.3 | NP_726856.1 | 9 - 10 | 5082 | 85 | ||
| NM_166969.2 | NP_726857.1 | 6 - 7 | 780 | 85 | ||
| NM_166970.2 | NP_726858.2 | 5 - 6 | 998 | 85 | ||
| NM_166971.2 | NP_726859.2 | 2 - 3 | 561 | 85 | ||
| GenBank mRNA | AY119511.1 | - | 2 - 3 | 561 | 85 | |
| BT133288.1 | - | 9 - 10 | 2485 | 85 | ||
More Information
Set Membership: |
|

dnc
Genomic Map