Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166961.2 | NP_726849.1 | 3 - 4 | 1620 | 56 | |
| NM_166962.2 | NP_726850.1 | 3 - 4 | 1617 | 56 | ||
| NM_166964.2 | NP_726852.2 | 3 - 4 | 1620 | 56 | ||
| NM_166965.2 | NP_726853.2 | 3 - 4 | 1617 | 56 | ||
More Information
Set Membership: |
|

dnc
Genomic Map