Assay Details
| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166968.3 | NP_726856.1 | 5 - 6 | 4513 | 92 | |
| NM_166970.2 | NP_726858.2 | 1 - 2 | 429 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dnc| Gene Symbol: dnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166968.3 | NP_726856.1 | 5 - 6 | 4513 | 92 | |
| NM_166970.2 | NP_726858.2 | 1 - 2 | 429 | 92 | ||
Set Membership: |
|