Assay Details
| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169180.1 | NP_001162651.1 | 5 - 6 | 2387 | 59 | |
| NM_001169181.3 | NP_001162652.1 | 4 - 5 | 1457 | 59 | ||
| NM_001169182.3 | NP_001162653.1 | 4 - 5 | 2312 | 59 | ||
| NM_001169183.2 | NP_001162654.1 | 4 - 5 | 1685 | 59 | ||
| NM_001258583.2 | NP_001245512.1 | 4 - 5 | 2312 | 59 | ||
| NM_001258584.2 | NP_001245513.1 | 4 - 5 | 1547 | 59 | ||
| NM_001297913.1 | NP_001284842.1 | 4 - 5 | 1949 | 59 | ||
| NM_130715.4 | NP_570071.3 | 4 - 5 | 2171 | 59 | ||
| NM_166983.3 | NP_726879.1 | 4 - 5 | 1547 | 59 | ||
| NM_166984.3 | NP_726880.1 | 4 - 5 | 1688 | 59 | ||
| GenBank mRNA | AY075354.1 | - | 3 - 4 | 1437 | 59 | |
More Information
Set Membership: |
|

Mnt
Genomic Map