Assay Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080324.4 | NP_525063.1 | 2 - 3 | 454 | 82 | |
| NM_166985.3 | NP_726881.1 | 2 - 3 | 454 | 82 | ||
| NM_166986.3 | NP_726882.1 | 3 - 4 | 650 | 82 | ||
| GenBank mRNA | AY024366.1 | - | 1 - 2 | 154 | 82 | |
| AY118912.1 | - | 2 - 3 | 421 | 82 | ||
| U23800.1 | - | 2 - 3 | 416 | 82 | ||
More Information
Set Membership: |
|

Rala
Genomic Map