Assay Details
| Gene Symbol: Psc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299439.1 | NP_001286368.1 | 3 - 4 | 1612 | 81 | |
| NM_001299440.1 | NP_001286369.1 | 3 - 4 | 1948 | 81 | ||
| NM_079001.3 | NP_523725.2 | 3 - 4 | 1948 | 81 | ||
| GenBank mRNA | BT100307.1 | - | 3 - 4 | 1914 | 81 | |
More Information
Set Membership: |
|

Psc
Genomic Map