Assay Details
| Gene Symbol: Su(z)2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169663.3 | NP_001163134.1 | 2 - 3 | 530 | 84 | |
| NM_001274004.1 | NP_001260933.1 | 2 - 3 | 530 | 84 | ||
| NM_079002.3 | NP_523726.2 | 2 - 3 | 309 | 84 | ||
More Information
Set Membership: |
|

Su(z)2
Genomic Map