Assay Details
| Gene Symbol: mRpS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079013.4 | NP_523737.1 | 1 - 2 | 227 | 68 | |
| GenBank mRNA | AY119210.1 | - | 1 - 2 | 167 | 68 | |
More Information
Set Membership: |
|

mRpS16
Genomic Map