Assay Details
| Gene Symbol: Sox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079015.3 | NP_523739.2 | 2 - 3 | 1814 | 66 | |
| GenBank mRNA | BT025204.1 | - | 2 - 3 | 1784 | 66 | |
More Information
Set Membership: |
|

Sox15
Genomic Map