Assay Details
| Gene Symbol: Hsc70-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299485.1 | NP_001286414.1 | 2 - 3 | 272 | 75 | |
| NM_079017.2 | NP_523741.2 | 2 - 3 | 403 | 75 | ||
| GenBank mRNA | AY118844.2 | - | 2 - 3 | 219 | 75 | |
More Information
Set Membership: |
|

Hsc70-5
Genomic Map