Assay Details
| Gene Symbol: Hsc70-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299485.1 | NP_001286414.1 | 3 - 4 | 853 | 61 | |
| NM_079017.2 | NP_523741.2 | 3 - 4 | 984 | 61 | ||
| GenBank mRNA | AY118844.2 | - | 3 - 4 | 800 | 61 | |
More Information
Set Membership: |
|

Hsc70-5
Genomic Map