Assay Details
| Gene Symbol: LamC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274045.1 | NP_001260974.1 | 2 - 3 | 1142 | 61 | |
| NM_079018.3 | NP_523742.2 | 2 - 3 | 1142 | 61 | ||
| GenBank mRNA | AY095046.1 | - | 2 - 3 | 1118 | 61 | |
More Information
Set Membership: |
|

LamC
Genomic Map