Assay Details
| Gene Symbol: LamC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079018.3 | NP_523742.2 | 3 - 4 | 1864 | 55 | |
| GenBank mRNA | AY095046.1 | - | 3 - 4 | 1840 | 55 | |
More Information
Set Membership: |
|

LamC
Genomic Map