Assay Details
| Gene Symbol: Jhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169695.2 | NP_001163166.1 | 4 - 5 | 620 | 64 | |
| NM_001299547.1 | NP_001286476.1 | 4 - 5 | 608 | 64 | ||
| NM_079034.5 | NP_523758.3 | 4 - 5 | 667 | 64 | ||
More Information
Set Membership: |
|

Jhe
Genomic Map