Assay Details
| Gene Symbol: Jhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169695.2 | NP_001163166.1 | 5 - 6 | 1363 | 114 | |
| NM_001299547.1 | NP_001286476.1 | 5 - 6 | 1351 | 114 | ||
| NM_079034.5 | NP_523758.3 | 5 - 6 | 1410 | 114 | ||
More Information
Set Membership: |
|

Jhe
Genomic Map