Assay Details
| Gene Symbol: Tsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079035.3 | NP_523759.1 | 1 - 2 | 241 | 49 | |
| GenBank mRNA | BT099700.1 | - | 1 - 2 | 216 | 49 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsf3| Gene Symbol: Tsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079035.3 | NP_523759.1 | 1 - 2 | 241 | 49 | |
| GenBank mRNA | BT099700.1 | - | 1 - 2 | 216 | 49 | |
Set Membership: |
|