Assay Details
| Gene Symbol: botv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079066.3 | NP_523790.1 | 1 - 2 | 332 | 76 | |
| GenBank mRNA | AB077850.1 | - | 1 - 2 | 324 | 76 | |
| AF132161.1 | - | 1 - 2 | 324 | 76 | ||
More Information
Set Membership: |
|

botv
Genomic Map