Assay Details
| Gene Symbol: Ssrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079106.3 | NP_523830.2 | 2 - 3 | 1772 | 157 | |
| GenBank mRNA | BT044339.1 | - | 2 - 3 | 1712 | 157 | |
| X68408.1 | - | 1753 | 157 | |||
More Information
Set Membership: |
|

Ssrp
Genomic Map