Assay Details
| Gene Symbol: slbo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299907.1 | NP_001286836.1 | 1 | 2421 | 99 | |
| NM_079119.3 | NP_523843.1 | 1 | 2626 | 99 | ||
| GenBank mRNA | BT009992.1 | - | 1 | 2625 | 99 | |
More Information
Set Membership: |
|

slbo
Genomic Map