Assay Details
| Gene Symbol: lov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014554.3 | NP_001014554.1 | 2 - 3 | 2627 | 63 | |
| NM_001299947.1 | NP_001286876.1 | 3 - 4 | 2732 | 63 | ||
| NM_079141.4 | NP_523865.2 | 2 - 3 | 2373 | 63 | ||
| NM_138150.4 | NP_611994.3 | 2 - 3 | 2373 | 63 | ||
| NM_145761.2 | NP_665704.1 | 2 - 3 | 2412 | 63 | ||
| GenBank mRNA | AY122101.2 | - | 3 - 4 | 2649 | 63 | |
More Information
Set Membership: |
|

lov
Genomic Map