Assay Details
| Gene Symbol: trh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043110.2 | NP_001036575.1 | 2 - 3 | 1136 | 81 | |
| NM_001103991.2 | NP_001097461.2 | 5 - 6 | 1152 | 81 | ||
| NM_001274278.1 | NP_001261207.1 | 2 - 3 | 1136 | 81 | ||
| NM_001274279.1 | NP_001261208.1 | 2 - 3 | 1136 | 81 | ||
| NM_001274280.1 | NP_001261209.1 | 5 - 6 | 1152 | 81 | ||
| NM_001274281.1 | NP_001261210.1 | 2 - 3 | 1136 | 81 | ||
| NM_079148.3 | NP_523872.2 | 2 - 3 | 1136 | 81 | ||
| GenBank mRNA | BT133348.1 | - | 2 - 3 | 1136 | 81 | |
More Information
Set Membership: |
|

trh
Genomic Map