Assay Details
| Gene Symbol: CkIIalpha-i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299961.1 | NP_001286890.1 | 6 - 7 | 1269 | 114 | |
| NM_079151.4 | NP_523875.2 | 6 - 7 | 1004 | 114 | ||
| NM_167840.3 | NP_728542.1 | 7 - 8 | 961 | 114 | ||
| GenBank mRNA | AY058626.1 | - | 6 - 7 | 968 | 114 | |
More Information
Set Membership: |
|

CkIIalpha-i3
Genomic Map