Assay Details
| Gene Symbol: Glut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103997.3 | NP_001097467.1 | 8 - 9 | 1867 | 64 | |
| NM_001103998.3 | NP_001097468.1 | 8 - 9 | 1888 | 64 | ||
| NM_001103999.4 | NP_001097469.2 | 10 - 11 | 2192 | 64 | ||
| NM_001104000.2 | NP_001097470.2 | 8 - 9 | 1867 | 64 | ||
| NM_001274301.1 | NP_001261230.1 | 8 - 9 | 1867 | 64 | ||
| NM_001274302.1 | NP_001261231.1 | 8 - 9 | 1867 | 64 | ||
| NM_001274303.1 | NP_001261232.1 | 11 - 12 | 2248 | 64 | ||
| NM_001274304.1 | NP_001261233.1 | 16 - 17 | 2979 | 64 | ||
| NM_001274307.1 | NP_001261236.1 | 13 - 14 | 2613 | 64 | ||
| NM_001274308.1 | NP_001261237.1 | 13 - 14 | 2613 | 64 | ||
| NM_001274309.1 | NP_001261238.1 | 13 - 14 | 2613 | 64 | ||
| NM_001299966.1 | NP_001286895.1 | 8 - 9 | 1867 | 64 | ||
| NM_001299967.1 | NP_001286896.1 | 8 - 9 | 1867 | 64 | ||
| NM_001316474.1 | NP_001303403.1 | 8 - 9 | 1867 | 64 | ||
| NM_079154.3 | NP_523878.1 | 8 - 9 | 1867 | 64 | ||
| NM_138229.2 | NP_612073.2 | 8 - 9 | 1867 | 64 | ||
| NM_167845.3 | NP_728557.1 | 8 - 9 | 1739 | 64 | ||
| GenBank mRNA | AF064703.2 | - | 8 - 9 | 1865 | 64 | |
| BT011383.1 | - | 8 - 9 | 1856 | 64 | ||
More Information
Set Membership: |
|

Glut1
Genomic Map