Assay Details
| Gene Symbol: rho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274326.1 | NP_001261255.1 | 2 - 3 | 673 | 58 | |
| NM_079159.3 | NP_523883.2 | 2 - 3 | 673 | 58 | ||
| GenBank mRNA | AY121641.1 | - | 2 - 3 | 649 | 58 | |
More Information
Set Membership: |
|

rho
Genomic Map