Assay Details
| Gene Symbol: dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079166.3 | NP_523890.2 | 3 - 4 | 1228 | 80 | |
| NM_167956.2 | NP_728757.1 | 4 - 5 | 2640 | 80 | ||
| GenBank mRNA | AY122244.1 | - | 4 - 5 | 2618 | 80 | |
| BT149883.1 | - | 3 - 4 | 1156 | 80 | ||
| BT150064.1 | - | 3 - 4 | 1193 | 80 | ||
More Information
Set Membership: |
|

dos
Genomic Map