Assay Details
| Gene Symbol: dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079166.3 | NP_523890.2 | 4 - 5 | 1345 | 118 | |
| NM_167956.2 | NP_728757.1 | 5 - 6 | 2757 | 118 | ||
| GenBank mRNA | AY122244.1 | - | 5 - 6 | 2735 | 118 | |
| BT149883.1 | - | 4 - 5 | 1273 | 118 | ||
| BT150064.1 | - | 4 - 5 | 1310 | 118 | ||
More Information
Set Membership: |
|

dos
Genomic Map