Assay Details
| Gene Symbol: Shab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202108.2 | NP_001189037.1 | 6 - 7 | 3059 | 56 | |
| NM_079170.6 | NP_523894.2 | 6 - 7 | 2957 | 56 | ||
More Information
Set Membership: |
|

Shab
Genomic Map